missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-formylbenzoate, 98+%

Product Code. 11441097
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11441097

Brand: Thermo Scientific Alfa Aesar B24091.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1571-08-0 | C9H8O3 | 164.16 g/mol

Methyl 4-formylbenzoate is used in the preparation of dimethyl terephthalate. It is also used as an active pharmaceutical ingredient intermediate and fluorescent brightener intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 4-formylbenzoate is used in the preparation of dimethyl terephthalate. It is also used as an active pharmaceutical ingredient intermediate and fluorescent brightener intermediate.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1571-08-0
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00006950
InChI Key FEIOASZZURHTHB-UHFFFAOYSA-N
Synonym methyl p-formylbenzoate, 4-carbomethoxybenzaldehyde, methyl terephthalaldehydate, 4-methoxycarbonyl benzaldehyde, benzoic acid, 4-formyl-, methyl ester, p-carbomethoxybenzaldehyde, methyl terephthaldehydate, methyl4-formylbenzoate, 4-formylbenzoic acid methyl ester, methyl-p-formyl benzoate
PubChem CID 15294
IUPAC Name methyl 4-formylbenzoate
SMILES COC(=O)C1=CC=C(C=O)C=C1

Specifications

Melting Point 59°C to 63°C
Density 1.2
Boiling Point 264°C to 266°C
Flash Point 128°C (262°F)
Quantity 100 g
Beilstein 775895
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 164.16
Percent Purity ≥98%
Chemical Name or Material Methyl 4-formylbenzoate
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 4-formylbenzoate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Methyl 4-formylbenzoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.