Learn More
Methotrexate, Thermo Scientific™
£102.65 - £548.00
Chemical Identifiers
| CAS | 59-05-2 |
|---|---|
| Molecular Formula | C20H22N8O5 |
| Molecular Weight (g/mol) | 454.447 |
| MDL Number | MFCD00150847 |
| InChI Key | FBOZXECLQNJBKD-ZDUSSCGKSA-N |
| Synonym | methotrexate, abitrexate, rheumatrex, amethopterin, mexate, metatrexan, hdmtx, trexall, methylaminopterinum, antifolan |
| PubChem CID | 126941 |
| ChEBI | CHEBI:44185 |
| IUPAC Name | (2S)-2-[[4-[(2,4-diaminopteridin-6-yl)methyl-methylamino]benzoyl]amino]pentanedioic acid |
| SMILES | CN(CC1=CN=C2C(=N1)C(=NC(=N2)N)N)C3=CC=C(C=C3)C(=O)NC(CCC(=O)O)C(=O)O |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15445349
|
Thermo Scientific Alfa Aesar
J63075.MC |
100mg |
£114.00 £102.65 / 100mg Save £11.35 9% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15435349
|
Thermo Scientific Alfa Aesar
J63075.03 |
1g |
£548.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
SolubilitySoluble in dilute HCl or dilute alkali hydroxide solutions (i.e. 0.1M NaOH). Insoluble in plain water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away from strong oxidizing agents. Protect from light
Chemical Identifiers
| 59-05-2 | |
| 454.447 | |
| FBOZXECLQNJBKD-ZDUSSCGKSA-N | |
| 126941 | |
| (2S)-2-[[4-[(2,4-diaminopteridin-6-yl)methyl-methylamino]benzoyl]amino]pentanedioic acid |
| C20H22N8O5 | |
| MFCD00150847 | |
| methotrexate, abitrexate, rheumatrex, amethopterin, mexate, metatrexan, hdmtx, trexall, methylaminopterinum, antifolan | |
| CHEBI:44185 | |
| CN(CC1=CN=C2C(=N1)C(=NC(=N2)N)N)C3=CC=C(C=C3)C(=O)NC(CCC(=O)O)C(=O)O |
Specifications
| 59-05-2 | |
| MFCD00150847 | |
| Hygroscopic; Light sensitive | |
| methotrexate, abitrexate, rheumatrex, amethopterin, mexate, metatrexan, hdmtx, trexall, methylaminopterinum, antifolan | |
| FBOZXECLQNJBKD-ZDUSSCGKSA-N | |
| (2S)-2-[[4-[(2,4-diaminopteridin-6-yl)methyl-methylamino]benzoyl]amino]pentanedioic acid | |
| 126941 | |
| 454.44 |
| C20H22N8O5 | |
| UN1544 | |
| 14,5985 | |
| Soluble in dilute HCl or dilute alkali hydroxide solutions (i;e; 0;1M NaOH); Insoluble in plain water. | |
| CN(CC1=CN=C2C(=N1)C(=NC(=N2)N)N)C3=CC=C(C=C3)C(=O)NC(CCC(=O)O)C(=O)O | |
| 454.447 | |
| CHEBI:44185 | |
| Methotrexate |
Safety and Handling
GHS H Statement
H301-H360-H315-H319
Toxic if swallowed.
May damage fertility or the unborn child.
Causes skin irritation.
Causes serious eye irritation.
P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
H301-H315-H319-H335-H340-H360FD
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ALKALOIDS, SOLID, N.O.S.
missing translation for 'einecsNumber' : 200-413-8
missing translation for 'rtecsNumber' : MA1225000
missing translation for 'tsca' : Yes
Recommended Storage : Store at -20°C
- Methotrexate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- Germ cell mutagenicity Category 1B
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H301-Toxic if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H340-May cause genetic defects.
- H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contain: Methotrexate
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.