missing translation for 'onlineSavingsMsg'
Learn More

Methotrexate, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar J63075.03

Quantity 1g

 View more versions of this product

Product Code. 15435349

  • £548.00 / 1g

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Soluble in dilute HCl or dilute alkali hydroxide solutions (i.e. 0.1M NaOH). Insoluble in plain water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away from strong oxidizing agents. Protect from light
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

59-05-2
454.447
FBOZXECLQNJBKD-ZDUSSCGKSA-N
126941
(2S)-2-[[4-[(2,4-diaminopteridin-6-yl)methyl-methylamino]benzoyl]amino]pentanedioic acid
C20H22N8O5
MFCD00150847
methotrexate, abitrexate, rheumatrex, amethopterin, mexate, metatrexan, hdmtx, trexall, methylaminopterinum, antifolan
CHEBI:44185
CN(CC1=CN=C2C(=N1)C(=NC(=N2)N)N)C3=CC=C(C=C3)C(=O)NC(CCC(=O)O)C(=O)O

Specifications

59-05-2
MFCD00150847
UN1544
14,5985
Soluble in dilute HCl or dilute alkali hydroxide solutions (i;e; 0;1M NaOH); Insoluble in plain water.
CN(CC1=CN=C2C(=N1)C(=NC(=N2)N)N)C3=CC=C(C=C3)C(=O)NC(CCC(=O)O)C(=O)O
454.447
CHEBI:44185
Methotrexate
C20H22N8O5
1g
Hygroscopic; Light sensitive
methotrexate, abitrexate, rheumatrex, amethopterin, mexate, metatrexan, hdmtx, trexall, methylaminopterinum, antifolan
FBOZXECLQNJBKD-ZDUSSCGKSA-N
(2S)-2-[[4-[(2,4-diaminopteridin-6-yl)methyl-methylamino]benzoyl]amino]pentanedioic acid
126941
454.44
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.