missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEF2B Mouse anti-Human, Clone: PCRP-MEF2B-2F9, Novus Biologicals™
Mouse Monoclonal Antibody
£216.00 - £415.00
Specifications
| Antigen | MEF2B |
|---|---|
| Clone | PCRP-MEF2B-2F9 |
| Concentration | 0.2 mg/ml |
| Dilution | Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunofluorescence 1-2 ug/ml, Immunoprecipitation 1-2 ug/100-500 ug protein, Protein Array |
| Applications | Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Peptide Array |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18351702
|
Bio-Techne
NBP3-13825-20UG |
20 μg |
£216.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18309026
|
Novus Biologicals
NBP3-13825-100UG |
100 μg |
£415.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MEF2B Monoclonal antibody specifically detects MEF2B in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Protein ArraySpecifications
| MEF2B | |
| 0.2 mg/ml | |
| Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Peptide Array | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q02080 | |
| 4207 | |
| IgG2a | |
| Protein A or G purified |
| PCRP-MEF2B-2F9 | |
| Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunofluorescence 1-2 ug/ml, Immunoprecipitation 1-2 ug/100-500 ug protein, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| LOC729991-MEF2B readthrough transcript, MEF2B, myocyte enhancer factor 2B | |
| Recombinant full-length human MEF2B protein (Uniprot: Q02080) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title