missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEF2B Mouse anti-Human, Clone: PCRP-MEF2B-2F9, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-13825-100UG
This item is not returnable.
View return policy
Description
MEF2B Monoclonal antibody specifically detects MEF2B in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Protein Array
Specifications
| MEF2B | |
| Monoclonal | |
| 0.2 mg/ml | |
| Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunofluorescence 1-2 ug/ml, Immunoprecipitation 1-2 ug/100-500 ug protein, Protein Array | |
| Q02080 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 4207 | |
| Store at 4°C. | |
| IgG2a |
| Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Peptide Array | |
| PCRP-MEF2B-2F9 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| LOC729991-MEF2B readthrough transcript, MEF2B, myocyte enhancer factor 2B | |
| Recombinant full-length human MEF2B protein (Uniprot: Q02080) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction