missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ MC500, MD600 Colorimeter Replacement Reagents: pH value
For use with MC500 and MD600 multi-parameter colorimeter
£10.90 - £21.00
Specifications
For Use With (Application) | Water quality testing |
---|---|
Storage Requirements | 20°C |
Test Method | Phenol red |
Product Code | Brand | Color | Format | pH Range | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Format | pH Range | Price | Quantity & Availability | |||
11312530
|
Lovibond™
511770BT |
Orange | Tablet | 5.1 |
£21.00
100 tablets |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||
10639973
|
Lovibond™
471040 |
Red | Liquid | 7.7 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Specifications
Water quality testing | |
20°C | |
Phenol red |