missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ MC500, MD600 Colorimeter Replacement Reagents: pH value

Product Code. 11312530
Change view
Click to view available options
Color:
Orange
Red
Format:
Liquid
Tablet
pH Range:
5.1
7.7
Unit Size:
100 tablets
15mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Format pH Range unitSize
11312530 Orange Tablet 5.1 100 tablets
10639973 Red Liquid 7.7 15mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11312530 Supplier Lovibond™ Supplier No. 511770BT

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For use with MC500 and MD600 multi-parameter colorimeter

Specifications

Color Orange
Composition Sodium Chloride 90 to 100%, non-hazardous components <2%
pH Range 5.1
Format Tablet
For Use With (Application) Water quality testing
For Use With (Equipment) MC500, MD600, SC450, MD100 pool control line, aquaPRO3 and aquaPRO6 photometers and scuba II electronic pool tester
No. of Tests 100
Storage Requirements 20°C
Test Method Phenol red
Type Phenol Red Photometer Tablet
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.