missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human MAPK13 (p38 delta), His Tag Recombinant Protein
Human MAPK13 (p38 delta), His Tag Recombinant Protein
£434.00 - £6900.00
Specifications
Accession Number | NP_002745 |
---|---|
Gene Alias | MAPK13 (p38 δ) |
KinaseFamily | MAP Kinase Family |
Species | Human |
Recombinant | Recombinant |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11593487
|
Invitrogen™
PV3656 |
10μg |
£434.00
10µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15448493
|
Invitrogen™
PR6835B |
100μg |
£1602.00
100µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10414842
|
Invitrogen™
PV4017 |
1 mg |
£6900.00
1mg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
MAPK13 (p38 delta) transduces a variety of extracellular signals, including cytokine and environmental stress responses.
Specifications
NP_002745 | |
MAP Kinase Family | |
Recombinant | |
Baculovirus |
MAPK13 (p38 δ) | |
Human | |
His Tag |