missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MAPK13 (p38 delta), His Tag Recombinant Protein

Human MAPK13 (p38 delta), His Tag Recombinant Protein

Brand:  Invitrogen™ PV4017

Code : 50

Additional Details : Weight : 0.00945kg

 View more versions of this product

Product Code. 10414842

  • £6900.00 / 1mg
Estimated Shipment: 04-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


For Research Use Only. All usage must comply with product instructions.
 

Description

Description

  • MAPK13 (p38 delta) is a serine/threonine kinase that transduces a variety of extracellular signals, including cytokine and environmental stress responses
  • MAPK13 also regulates the differentiation of keratinocytes
Specifications

Specifications

NP_002745
46.2 kDa
MAPK13 (p38 δ)
Human
His Tag
Ser/Thr Kinases: CMGC Group
Baculovirus
Frozen
Kinase Assay
1 mg
MAP Kinase Family
Recombinant
Storage Buffer: 50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Dry Ice
Full Length
Drug Development
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers