missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MAPK13 (p38 delta), His Tag Recombinant Protein

Human MAPK13 (p38 delta), His Tag Recombinant Protein

Brand:  Invitrogen™ PV3656

Code : 50

Additional Details : Weight : 0.05250kg

 View more versions of this product

Product Code. 11593487

  • £434.00 / 10µg
Estimated Shipment: 04-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


For Research Use Only. All usage must comply with product instructions.
 

Description

Description

MAPK13 (p38 delta) transduces a variety of extracellular signals, including cytokine and environmental stress responses.

Specifications

Specifications

NP_002745
5603
10μg
MAP Kinase Family
Recombinant
Storage Buffer: 50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Ser⁄Thr Kinases: CMGC Group
LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™, Omnia™ Bulk Substrate
mitogen-activated protein kinase 13
Kinases & Inhibitors
Liquid
Signal Transduction
Kinase Assay
46.2 kDa
MAPK13 (p38 δ)
Human
His Tag
Biochemical terbium phospho-specific activity, Biochemical phospho-specific activity (no antibody)
Dry Ice
MAPK13
Baculovirus
Recombinant, Full Length
Serine/Threonine Kinases
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers