missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LOC123688 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | LOC123688 |
|---|---|
| Concentration | 0.3 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16864595
|
Proteintech
26070-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16854595
|
Proteintech
26070-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Catalyzes the GTP-dependent phosphorylation of 5- hydroxy-L-lysine.Specifications
| LOC123688 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human | |
| A2RU49, Q5U5V2 | |
| 123688, 235386 | |
| AGPHD1 Fusion Protein Ag23420 | |
| Primary | |
| -20°C | |
| HYKK |
| 0.3 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| 5-hydroxy-L-lysine kinase, AGPHD1, Hydroxylysine kinase, HYKK | |
| Hykk | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title