missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LOC123688 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26070-1-AP-20UL
This item is not returnable.
View return policy
Description
Catalyzes the GTP-dependent phosphorylation of 5- hydroxy-L-lysine.Specifications
| LOC123688 | |
| Polyclonal | |
| Unconjugated | |
| HYKK | |
| 5-hydroxy-L-lysine kinase, AGPHD1, Hydroxylysine kinase, HYKK | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 123688, 235386 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| A2RU49, Q5U5V2 | |
| Hykk | |
| AGPHD1 Fusion Protein Ag23420 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction