Learn More
LOC123688 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26070-1-AP-150UL
 View more versions of this product
Product Code. 16854595
  • £349.00 / 150µL
Description
Catalyzes the GTP-dependent phosphorylation of 5- hydroxy-L-lysine.
Specifications
Specifications
LOC123688
Polyclonal
Unconjugated
HYKK
5-hydroxy-L-lysine kinase, AGPHD1, Hydroxylysine kinase, HYKK
Rabbit
Antigen Affinity Chromatography
RUO
123688, 235386
-20°C
Liquid
Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
A2RU49, Q5U5V2
Hykk
AGPHD1 Fusion Protein Ag23420
150 μL
Primary
Human, Mouse