missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LMF2 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
£161.00 - £341.00
Specifications
| Antigen | LMF2 |
|---|---|
| Concentration | 0.5 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17979502
![]() |
Invitrogen™
PA5145467 |
200 μL |
£341.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17938842
|
Invitrogen™
PA5145468 |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Involved in the maturation of specific proteins in the endoplasmic reticulum. May be required for maturation and transport of active lipoprotein lipase (LPL) through the secretory pathway.Specifications
| LMF2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Pig | |
| Q9BU23 | |
| 100518488, 91289 | |
| Recombinant Human LMF2 protein, Leu454-Lys707 (Accession #Q9BU23). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| 0.5 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| AI451006; Lipase maturation factor 2; LMF2; RGD1306274; TMEM112B; Tmem153; Transmembrane protein 112B; transmembrane protein 153 | |
| LMF2 | |
| IgG | |
| Antigen affinity chromatography, Protein A | |
| LMF2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title