missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LMF2 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5145468

 View more versions of this product

Product Code. 17938842

  • £161.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Involved in the maturation of specific proteins in the endoplasmic reticulum. May be required for maturation and transport of active lipoprotein lipase (LPL) through the secretory pathway.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LMF2
Polyclonal
Unconjugated
LMF2
AI451006; Lipase maturation factor 2; LMF2; RGD1306274; TMEM112B; Tmem153; Transmembrane protein 112B; transmembrane protein 153
Rabbit
Antigen affinity chromatography, Protein A
RUO
100518488, 91289
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.5 mg/mL
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Q9BU23
LMF2
Recombinant Human LMF2 protein, Leu454-Lys707 (Accession #Q9BU23).
20 μL
Primary
Human, Pig
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.