missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LMF2 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5145467
This item is not returnable.
View return policy
Description
Involved in the maturation of specific proteins in the endoplasmic reticulum. May be required for maturation and transport of active lipoprotein lipase (LPL) through the secretory pathway.
Specifications
| LMF2 | |
| Polyclonal | |
| Unconjugated | |
| LMF2 | |
| AI451006; Lipase maturation factor 2; LMF2; RGD1306274; TMEM112B; Tmem153; Transmembrane protein 112B; transmembrane protein 153 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 100518488, 91289 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| Q9BU23 | |
| LMF2 | |
| Recombinant Human LMF2 protein, Leu454-Lys707 (Accession #Q9BU23). | |
| 200 μL | |
| Primary | |
| Human, Pig | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction