missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIV-1/Zip6 Antibody (13F1), Novus Biologicals™
Human Monoclonal Antibody
£269.00 - £431.00
Specifications
| Antigen | LIV-1/Zip6 |
|---|---|
| Clone | 13F1 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30033314
|
Novus Biologicals
NBP3-26316-50ul |
50 μL |
£269.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30036407
|
Novus Biologicals
NBP3-26316-100ul |
100 μL |
£431.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LIV-1/Zip6 Monoclonal antibody specifically detects LIV-1/Zip6 in Human samples. It is validated for ELISASpecifications
| LIV-1/Zip6 | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| zinc transporter ZIP6, LIV-1, solute carrier family 39 (zinc transporter), member 6 | |
| Recombinant Human LIV-1/Zip6 protein [UniProt Q13433] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 13F1 | |
| ELISA | |
| Unconjugated | |
| Human | |
| Cancer, Endocrinology, Signal Transduction | |
| 50% Glycerol, 0.01M PBS, PH 7.4 | |
| 25800 | |
| IgG1 | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title