missing translation for 'onlineSavingsMsg'
Learn More

LIV-1/Zip6 Antibody (13F1), Novus Biologicals™

Human Monoclonal Antibody

Brand:  Novus Biologicals NBP3-26316-50ul

 View more versions of this product

Product Code. 30033314

  • £269.00 / 50µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

LIV-1/Zip6 Monoclonal antibody specifically detects LIV-1/Zip6 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LIV-1/Zip6
Monoclonal
Unconjugated
50% Glycerol, 0.01M PBS, PH 7.4
Human
Affinity purified
RUO
Primary
Human
Purified
ELISA
13F1
ELISA
zinc transporter ZIP6, LIV-1, solute carrier family 39 (zinc transporter), member 6
Recombinant Human LIV-1/Zip6 protein [UniProt Q13433]
50 μL
Cancer, Endocrinology, Signal Transduction
25800
Store at -20 to -70C. Avoid freeze-thaw cycles.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.