missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIV-1/Zip6 Antibody (13F1), Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-26316-100ul
This item is not returnable.
View return policy
Description
LIV-1/Zip6 Monoclonal antibody specifically detects LIV-1/Zip6 in Human samples. It is validated for ELISA
Specifications
| LIV-1/Zip6 | |
| Monoclonal | |
| Unconjugated | |
| 50% Glycerol, 0.01M PBS, PH 7.4 | |
| Human | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 13F1 | |
| ELISA | |
| zinc transporter ZIP6, LIV-1, solute carrier family 39 (zinc transporter), member 6 | |
| Recombinant Human LIV-1/Zip6 protein [UniProt Q13433] | |
| 100 μL | |
| Cancer, Endocrinology, Signal Transduction | |
| 25800 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction