missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LC3A Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£171.00 - £368.00
Specifications
| Antigen | LC3A |
|---|---|
| Concentration | 1.09 mg/mL |
| Dilution | Western Blot 1:1000, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 1:75, Immunohistochemistry-Paraffin 1:400 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18237226
|
Novus Biologicals
NBP1-78964SS |
0.025 mL |
£171.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18726583
|
Novus Biologicals
NBP1-78964 |
£368.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Descripción
LC3A Polyclonal specifically detects LC3A in Human, Mouse, Rat, Porcine, Bovine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Especificaciones
| LC3A | |
| Western Blot 1:1000, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 1:75, Immunohistochemistry-Paraffin 1:400 | |
| Polyclonal | |
| Rabbit | |
| Macroautophagy | |
| PBS and 30% Glycerol with 0.05% Sodium Azide | |
| ATG8E, Autophagy-related protein LC3 A, Autophagy-related ubiquitin-like modifier LC3 A, LC3, LC3A, lc3-i, ii, MAP1A/1B light chain 3 A, MAP1ALC3MAP1A/MAP1B LC3 A, MAP1BLC3MAP1A/MAP1B light chain 3 A, microtubule-associated protein 1 light chain 3 alphaMAP1 light chain 3-like protein 1, microtubule-associated proteins 1A/1B light chain 3, microtubule-associated proteins 1A/1B light chain 3A | |
| MAP1LC3B | |
| IgG | |
| Affinity Purified | |
| This LC3I antibody specifically detects the cytosolic form of LC3 before it gets converted into LC3II during autophagy. |
| 1.09 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat, Pig, Bovine, Primate | |
| Q9H492 | |
| 84557 | |
| A synthetic peptide made to a C-terminal region of the human LC3 protein (within residues 50-120). [Swiss-Prot Q9H492] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto