Learn More
LC3A Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-78964
This item is not returnable.
View return policy
Description
LC3A Polyclonal specifically detects LC3A in Human, Mouse, Rat, Porcine, Bovine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Polyclonal | |
| Human, Rat, Pig | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |