missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LC3A Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-78964SS
This item is not returnable.
View return policy
Description
LC3A Polyclonal specifically detects LC3A in Human, Mouse, Rat, Porcine, Bovine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| LC3A | |
| Polyclonal | |
| Unconjugated | |
| PBS and 30% Glycerol with 0.05% Sodium Azide | |
| ATG8E, Autophagy-related protein LC3 A, Autophagy-related ubiquitin-like modifier LC3 A, LC3, LC3A, lc3-i, ii, MAP1A/1B light chain 3 A, MAP1ALC3MAP1A/MAP1B LC3 A, MAP1BLC3MAP1A/MAP1B light chain 3 A, microtubule-associated protein 1 light chain 3 alphaMAP1 light chain 3-like protein 1, microtubule-associated proteins 1A/1B light chain 3, microtubule-associated proteins 1A/1B light chain 3A | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This LC3I antibody specifically detects the cytosolic form of LC3 before it gets converted into LC3II during autophagy. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.09 mg/mL | |
| Western Blot 1:1000, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 1:75, Immunohistochemistry-Paraffin 1:400 | |
| Q9H492 | |
| MAP1LC3B | |
| A synthetic peptide made to a C-terminal region of the human LC3 protein (within residues 50-120). [Swiss-Prot Q9H492] | |
| 0.025 mL | |
| Macroautophagy | |
| 84557 | |
| Human, Mouse, Rat, Pig, Bovine, Primate | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction