missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LATS1 Mouse anti-Human, Clone: 2A8F1, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | LATS1 |
|---|---|
| Clone | 2A8F1 |
| Concentration | 0.7 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16868565
|
Proteintech
66569-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16858565
|
Proteintech
66569-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Plays a role in maintece of the normal epithelial architecture through the repression of SNAI1 transcription in a histone deacetylase-dependent manner, and thus the regulation of E-cadherin levels.Specifications
| LATS1 | |
| 0.7 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| h warts, LATS1, WARTS, WARTS protein kinase, wts | |
| LATS1 | |
| IgG1 | |
| Protein G | |
| Antibody |
| 2A8F1 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| O95835 | |
| 9113 | |
| LATS1 Fusion Protein Ag11140 | |
| Primary | |
| -20°C | |
| LATS1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title