missing translation for 'onlineSavingsMsg'
Learn More

LATS1 Mouse anti-Human, Clone: 2A8F1, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66569-1-IG-20UL

 View more versions of this product

Product Code. 16868565

  • £135.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Plays a role in maintece of the normal epithelial architecture through the repression of SNAI1 transcription in a histone deacetylase-dependent manner, and thus the regulation of E-cadherin levels.
Specifications

Specifications

LATS1
Monoclonal
0.7 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O95835
LATS1
LATS1 Fusion Protein Ag11140
20 μL
Primary
Human
Antibody
IgG1
Western Blot
2A8F1
Unconjugated
LATS1
h warts, LATS1, WARTS, WARTS protein kinase, wts
Mouse
Protein G
RUO
9113
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.