missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LATS1 Mouse anti-Human, Clone: 2A8F1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66569-1-IG-150UL
This item is not returnable.
View return policy
Description
Plays a role in maintece of the normal epithelial architecture through the repression of SNAI1 transcription in a histone deacetylase-dependent manner, and thus the regulation of E-cadherin levels.Specifications
| LATS1 | |
| Monoclonal | |
| 0.7 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O95835 | |
| LATS1 | |
| LATS1 Fusion Protein Ag11140 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 2A8F1 | |
| Unconjugated | |
| LATS1 | |
| h warts, LATS1, WARTS, WARTS protein kinase, wts | |
| Mouse | |
| Protein G | |
| RUO | |
| 9113 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction