Learn More
Thermo Scientific™ LanthaScreen™ Tb-pSTAT3 [pTyr705] Antibody Kit
LanthaScreen™ Tb-pSTAT3 [pTyr705] Antibody Kit
£3965.00 - £19140.00
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
10227953
|
Thermo Scientific™
PV4846 |
25 μg |
£3965.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
10318914
|
Thermo Scientific™
PV4847 |
1 mg |
£19140.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
A part of the JAK/STAT Pathway, STAT3 (Signal Transducer and Activator of Transcription 3) is phosphorylated at Tyr705 by the Janus family of tyrosine kinases.For use in combination with the GFP-STAT3 physiological protein substrate (PV5212) and the LanthaScreen™ STAT3 GripTite™ cell line (K1488) to evaluate inhibitors in medium to high throughput screening applications.