Learn More
Thermo Scientific™ LanthaScreen™ Tb-pSTAT3 [pTyr705] Antibody Kit
LanthaScreen™ Tb-pSTAT3 [pTyr705] Antibody Kit
Brand: Thermo Scientific™ PV4847
This item is not returnable.
View return policy
Description
- A part of the JAK/STAT Pathway
- STAT3 (Signal Transducer and Activator of Transcription 3) is phosphorylated at Tyr705 by the Janus family of tyrosine kinases
- For use in combination with the GFP-STAT3 physiological protein substrate (PV5212) and the LanthaScreen™ STAT3 GripTite™ cell line (K1488)
- For inhibitor evaluation in medium to high throughput screening applications
Specifications
| Tb (Terbium)/GFP (EmGFP) | |
| Terbium-labeled anti-STAT3 [pTyr705] antibody in HEPES buffered saline. Store at -20°C. | |
| 340⁄490, 520 | |
| Dry Ice | |
| Hu | |
| TR-FRET |
| 1 mg | |
| 384-well Plate | |
| Human | |
| End Point | |
| Tyrosine Kinases | |
| JAK1, JAK2, JAK3, TYK2 |