Learn More
Thermo Scientific™ LanthaScreen™ Tb-pMAP2K1 (pSer217/221) Antibody Kit
LanthaScreen™ Tb-pMAP2K1 (pSer217/221) Antibody Kit
£3535.00 - £14930.00
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
10206313
|
Thermo Scientific™
PV4813 |
25 μg |
£3535.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
10368273
|
Thermo Scientific™
PV4814 |
1 mg |
£14930.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
A member of the MAP Kinase family, MAP2K1 (mitogen-activated protein kinase kinase 1) is phosphorylated at Ser217/221 by a range of MAPK family members including Raf and B-Raf. For use in combination with the Fluorescein-MAP2K1 physiological protein substrate (PV4812) to evaluate inhibitors in medium to high throughput screening applications.For Research Use Only. Not for use in diagnostic procedures.