missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ LanthaScreen™ Tb-pMAP2K1 (pSer217/221) Antibody Kit
LanthaScreen™ Tb-pMAP2K1 (pSer217/221) Antibody Kit
Brand: Thermo Scientific™ PV4814
This item is not returnable.
View return policy
Description
- A member of the MAP Kinase family
- MAP2K1 (mitogen-activated protein kinase kinase 1) is phosphorylated at Ser217/221 by a range of MAPK family members including Raf and B-Raf
- For use in combination with the Fluorescein-MAP2K1 physiological protein substrate (PV4812) to evaluate inhibitors in medium to high throughput screening applications
Specifications
| Tb (Terbium)/FITC | |
| 10454, 10746, 1326, 4215, 4217, 4293, 4294, 4296, 5894, 673, 6885 | |
| 384-well Plate | |
| Human | |
| End Point | |
| Serine/Threonine Kinases | |
| BRAF, MAP3K2, MAP3K3, MAP3K5, MAP3K7, MAP3K8, MAP3K9, MAP3K10, MAP3K11 |
| 1 mg | |
| Terbium-labeled anti-MAP2K1 [pSer217/221] antibody in HEPES buffered saline. Store at -20°C. | |
| 340/490, 520 | |
| Dry Ice | |
| Hu | |
| TR-FRET |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction