missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Lactyl Lysine Antibody Cocktail, Invitrogen™

Mouse Cocktail Antibody

£380.00 - £570.00

Specifications

Antigen Lactyl Lysine
Concentration 1 mg/mL
Applications Western Blot
Classification Cocktail
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30201768
View Documents
Invitrogen™
MA556375
200 μL
£570.00
200µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30195745
View Documents
Invitrogen™
MA555615
50 μL
£380.00
50µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Lactyl Lysine Cocktail antibody specifically detects Lactyl Lysine in Chemical samples. It is validated for Western Blot
Specifications

Specifications

Lactyl Lysine
Western Blot
Unconjugated
Mouse
Chemical
l-Lactyl-lysine; Lysine l-lactylation
IgG
Affinity chromatography
Antibody
1 mg/mL
Cocktail
Liquid
RUO
PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Lactyl Lysine peptide.
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.