missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Lactyl Lysine Antibody Cocktail, Invitrogen™

Mouse Cocktail Antibody

Brand:  Invitrogen™ MA556375

 View more versions of this product

Product Code. 30201768

  • £570.00 / 200µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Lactyl lysine is derivative of the amino acid residue lysine where the sidechain ammonium group undergoes lactylation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Lactyl Lysine
Cocktail
Unconjugated
l-Lactyl-lysine; Lysine l-lactylation
Lactyl Lysine peptide.
200 μL
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
1 mg/mL
PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Mouse
Affinity chromatography
RUO
Chemical
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.