missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Lactyl Lysine Antibody Cocktail, Invitrogen™
Mouse Cocktail Antibody
Brand: Invitrogen™ MA555615
This item is not returnable.
View return policy
Description
Lactyl lysine is derivative of the amino acid residue lysine where the sidechain ammonium group undergoes lactylation.
Specifications
| Lactyl Lysine | |
| Cocktail | |
| Unconjugated | |
| l-Lactyl-lysine; Lysine l-lactylation | |
| Lactyl Lysine peptide. | |
| 50 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction