missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labconco™ VACUUBRAND™ Hybrid™ Vacuum Pumps
Integrates high performance rotary vane pump with a chemical-resistant diaphragm pump to create a low maintenance, longer-lasting pump
£7920.00
Specifications
Weight (English) | 66 lb. |
---|---|
Weight (Metric) | 30 kg |
Overall Height (English) | 15 in. |
Overall Height (Metric) | 38.1 cm |
Overall Length (English) | 9 in. |
Description
Designed for use with acids and other chemicals including TFA, TFA by-products, acetonitrile, HBr and HNO3, present in samples such as HPLC-prepared and peptide purified materials.
- Combination pump system consists of a two-stage rotary vane pump and two-stage, chemically-resistant diaphragm pump
- Pressure control valve compensates for the different volumes displaced by the two pumps
- Ultimate vacuum (partial pressure) 3 × 10(-3)mBar (2.25 micron)
- Ready to use and fully charged with vacuum pump oil (approximately 1L)
- 0.75 in. OD inlet adapter
- Refill pump oil sold separately, 10-030-181
Specifications
66 lb. | |
15 in. | |
9 in. | |
18.9 in. |
30 kg | |
38.1 cm | |
22.9 cm | |
Vacuum Pump |