missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ VACUUBRAND™ Hybrid™ Vacuum Pumps

Integrates high performance rotary vane pump with a chemical-resistant diaphragm pump to create a low maintenance, longer-lasting pump

Brand:  Labconco™ 7584002


Code : 50

Additional Details : Weight : 30.00000kg

 View more versions of this product

Product Code. 14801282

  • £7920.00 / Each
Estimated Shipment: 03-10-2024
to see stock.
Add to basket

Description

Description

Designed for use with acids and other chemicals including TFA, TFA by-products, acetonitrile, HBr and HNO3, present in samples such as HPLC-prepared and peptide purified materials.

  • Combination pump system consists of a two-stage rotary vane pump and two-stage, chemically-resistant diaphragm pump
  • Pressure control valve compensates for the different volumes displaced by the two pumps
  • Ultimate vacuum (partial pressure) 3 × 10(-3)mBar (2.25 micron)
  • Ready to use and fully charged with vacuum pump oil (approximately 1L)
  • 0.75 in. OD inlet adapter
  • Refill pump oil sold separately, 10-030-181

Specifications

Specifications

1.6 A
230 V 50/60 Hz 1.6 A
30 kg
15 in.
9 in.
18.9 in.
230 V
1L bottle of vacuum pump oil, power switch, power cord and plug
66 lb.
50/60 Hz
38.1 cm
22.9 cm
Vacuum Pump
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers