missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ XPA Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Provide accurate, reproducible results with the Epredia™ XPA Ab-1, Mouse Monoclonal Antibody.

£120.36 - £545.48

Specifications

Antigen XPA Ab-1
Clone 12F5
Applications Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 6
Product Code Brand Quantity Form Concentration Price Quantity & Availability  
Product Code Brand Quantity Form Concentration Price Quantity & Availability  
12621917
View Documents
Epredia™
MS-650-P0
100 μL Concentrated 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12641917
View Documents
Epredia™
MS-650-P1ABX
100 μL Concentrated 1mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12651917
View Documents
Epredia™
MS-650-PABX
200 μL Concentrated 1mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12631917
View Documents
Epredia™
MS-650-P1
500 μL Concentrated 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12611917
View Documents
Epredia™
MS-650-P
1 mL Concentrated 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12691917
View Documents
Epredia™
MS-650-R7
7 mL Prediluted Prediluted This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

The XPA (xeroderma pigmentosum group A) protein specifically recognizes the UV-or chemically damaged DNA lesions, and triggers the nucleotide excision repair process. XPA binds to the replication protein A (RPA) or the excision repair cross complementing 1 protein (ERCC1). In the absence of nucleotide excision repair persisting (unrepaired) DNA lesions (adducts) may lead to the accumulation of gene mutations and ultimately to cancer. Xeroderma pigmentosum patients have a > 2000 fold increased risk to develop skin cancer at sun-exposed areas.

Host Species: Mouse

Clone: 12F5

Isotype: IgG2a/k

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Recombinant human XPA protein

Molecular Weight: ∽40kDa

Positive Control: Ls174T or MCF-7 cells or human tonsil

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

XPA Ab-1
Western Blot
Unconjugated
RUO
Human
IgG2a κ
12F5
Monoclonal
Mouse
DNA Repair
Recombinant human XPA protein
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.