missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ p504s/AMACR, Rabbit Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ p504s/AMACR, Rabbit Monoclonal Antibody.

£224.00 - £1177.95

Specifications

Antigen p504s/AMACR
Clone 13H4
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 5
Product Code Brand Quantity Form Price Quantity & Availability  
Product Code Brand Quantity Form Price Quantity & Availability  
12643787
View Documents
Epredia™
RM-9130-A0
100 μL Concentrated This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12623787
View Documents
Epredia™
RM-9130-A1
500 μL Concentrated This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12613787
View Documents
Epredia™
RM-9130-A
1 mL Concentrated
ONLINE EXCLUSIVE PRICE
£1255.00
£1177.95 / 1mL
Save £77.05
6% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12663787
View Documents
Epredia™
RM-9130-R7
7 mL Prediluted
ONLINE EXCLUSIVE PRICE
£290.00
£272.81 / 7mL
Save £17.19
5% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12673787
View Documents
Epredia™
RM-9130-RQ
12 mL Ready-to-Use
ONLINE EXCLUSIVE PRICE
£381.00
£357.71 / 12mL
Save £23.29
6% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Alpha methylacyl-CoA-racemase (AMACR), also known as p504s is an enzyme involved in the metabolism of branched-chain fatty acids. p504s is expressed in various normal tissues and carcinomas, while it is overexpressed over 30-fold in prostate carcinoma compared to benign glandular tissues1. When p504s is overexpressed in atypical glands without basal cells, it establishes a diagnosis of prostatic carcinoma. High-grade prostatic intraepithelial neoplasia, atypical adenomatous hyperplasia and urothelial carcinoma may also express p504s.

Host Species: Rabbit

Clone: 13H4

Isotype: IgG

Species Reactivity: Human. Others not tested.

Immunogen: Human p504s/AMACR polypeptide

Molecular Weight: ∽45 kDa

Positive Control: Prostate carcinoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Specifications

p504s/AMACR
Immunohistochemistry (Paraffin)
Unconjugated
ASR
Human p504s/AMACR polypeptide
13H4
Monoclonal
Rabbit
Cancer and Tumor Biology
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.