missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ p504s/AMACR, Rabbit Monoclonal Antibody

Product Code. 12613787 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
Unit Size:
0.1mL
12mL
1mL
500µL
7mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Form unitSize
12613787 1 mL Concentrated 1mL
12643787 100 μL Concentrated 0.1mL
12623787 500 μL Concentrated 500µL
12663787 7 mL Prediluted 7mL
12673787 12 mL Ready-to-Use 12mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 12613787 Supplier Epredia™ Supplier No. RM9130A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ p504s/AMACR, Rabbit Monoclonal Antibody.

Alpha methylacyl-CoA-racemase (AMACR), also known as p504s is an enzyme involved in the metabolism of branched-chain fatty acids. p504s is expressed in various normal tissues and carcinomas, while it is overexpressed over 30-fold in prostate carcinoma compared to benign glandular tissues1. When p504s is overexpressed in atypical glands without basal cells, it establishes a diagnosis of prostatic carcinoma. High-grade prostatic intraepithelial neoplasia, atypical adenomatous hyperplasia and urothelial carcinoma may also express p504s.

Host Species: Rabbit

Clone: 13H4

Isotype: IgG

Species Reactivity: Human. Others not tested.

Immunogen: Human p504s/AMACR polypeptide

Molecular Weight: ∽45 kDa

Positive Control: Prostate carcinoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen p504s/AMACR
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 13H4
Conjugate Unconjugated
Host Species Rabbit
Immunogen Human p504s/AMACR polypeptide
Quantity 1 mL
Regulatory Status ASR
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.