missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ p504s/AMACR, Rabbit Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ p504s/AMACR, Rabbit Monoclonal Antibody.

Brand:  Epredia™ RM-9130-A0

 View more versions of this product

Product Code. 12643787

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Alpha methylacyl-CoA-racemase (AMACR), also known as p504s is an enzyme involved in the metabolism of branched-chain fatty acids. p504s is expressed in various normal tissues and carcinomas, while it is overexpressed over 30-fold in prostate carcinoma compared to benign glandular tissues1. When p504s is overexpressed in atypical glands without basal cells, it establishes a diagnosis of prostatic carcinoma. High-grade prostatic intraepithelial neoplasia, atypical adenomatous hyperplasia and urothelial carcinoma may also express p504s.

Host Species: Rabbit

Clone: 13H4

Isotype: IgG

Species Reactivity: Human. Others not tested.

Immunogen: Human p504s/AMACR polypeptide

Molecular Weight: ∽45 kDa

Positive Control: Prostate carcinoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

p504s/AMACR
Monoclonal
Unconjugated
Human p504s/AMACR polypeptide
ASR
Primary
Concentrated
Immunohistochemistry (Paraffin)
13H4
Rabbit
100 μL
Cancer and Tumor Biology
Human
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.