missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Mast Cell Tryptase Ab-2 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Epredia™ Mast Cell Tryptase Ab-2, Mouse Monoclonal Antibody provides accurate, reproducible results.

£119.34 - £530.40

Specifications

Antigen Mast Cell Tryptase Ab-2
Clone AA1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 6
Product Code Brand Quantity Form Concentration Price Quantity & Availability  
Product Code Brand Quantity Form Concentration Price Quantity & Availability  
12639296
View Documents
Epredia™
MS-1216-P0
100 μL Concentrated 50μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12659296
View Documents
Epredia™
MS-1216-P1ABX
100 μL Concentrated 1mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12669296
View Documents
Epredia™
MS-1216-PABX
200 μL Concentrated 1mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12649296
View Documents
Epredia™
MS-1216-P1
500 μL Concentrated 50μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12629296
View Documents
Epredia™
MS-1216-P
1 mL Concentrated 50μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12699296
View Documents
Epredia™
MS-1216-R7
7 mL Prediluted
ONLINE EXCLUSIVE PRICE
£342.00
£320.59 / 7mL
Save £21.41
6% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Mast cells contain a number of preformed chemical mediators such as histamine, chymase, carboxypeptidase and proteolytic tryptase. A substantial quantity of tryptase is estimated to be found in mast cells of skin and lung and suggestes this enzyme plays a major role in mast cell mediated events. In vitro studies indicate tryptase can cleave C3 to form C3a anaphylatoxin, inactivate fibrinogen as a coaguable substrate for thrombin and activate latent collagenase. Human mast cell tryptase has considerable potential as a marker for mast cell activation as well as being a mediator of inflammation.

Host Species: Mouse

Clone: AA1

Isotype: IgG1

Species Reactivity: Human, Monkey, Dog and Cat. Others not known.

Epitope: Not determined

Immunogen: Human mast cell tryptase purified from human lung tissues

Positive Control: Tonsil

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Mast Cell Tryptase Ab-2
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
RUO
Canine, Feline, Human, Non-human Primate
IgG1
AA1
Monoclonal
Mouse
Enzyme
Human mast cell tryptase purified from human lung tissues
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.