missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Mast Cell Tryptase Ab-2 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Epredia™ Mast Cell Tryptase Ab-2, Mouse Monoclonal Antibody provides accurate, reproducible results.

Brand:  Epredia™ MS-1216-P1ABX

 View more versions of this product

Product Code. 12659296

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Mast cells contain a number of preformed chemical mediators such as histamine, chymase, carboxypeptidase and proteolytic tryptase. A substantial quantity of tryptase is estimated to be found in mast cells of skin and lung and suggestes this enzyme plays a major role in mast cell mediated events. In vitro studies indicate tryptase can cleave C3 to form C3a anaphylatoxin, inactivate fibrinogen as a coaguable substrate for thrombin and activate latent collagenase. Human mast cell tryptase has considerable potential as a marker for mast cell activation as well as being a mediator of inflammation.

Host Species: Mouse

Clone: AA1

Isotype: IgG1

Species Reactivity: Human, Monkey, Dog and Cat. Others not known.

Epitope: Not determined

Immunogen: Human mast cell tryptase purified from human lung tissues

Positive Control: Tonsil

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Mast Cell Tryptase Ab-2
Monoclonal
1mg/mL
Mouse
100 μL
Enzyme
Canine, Feline, Human, Non-human Primate
IgG1
Immunohistochemistry (Paraffin), Western Blot
AA1
Unconjugated
Human mast cell tryptase purified from human lung tissues
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.