Learn More
Cytokeratin 7 Ab-2 Mouse Monoclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Cytokeratin 7 Ab-2, Mouse Monoclonal Antibody.
£98.33 - £412.08
Specifications
| Antigen | Cytokeratin 7 Ab-2 |
|---|---|
| Clone | OV-TL 12/30 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Form | Concentration | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Form | Concentration | Price | Quantity & Availability | |||
|
12649636
|
Epredia
MS-1352-P0 |
100 μL | Concentrated | 200μg/mL | |||||
|
12669636
|
Epredia
MS-1352-P1ABX |
100 μL | Concentrated | 1mg/mL | |||||
|
12679636
|
Epredia
MS-1352-PABX |
200 μL | Concentrated | 1mg/mL | |||||
|
12659636
|
Epredia
MS-1352-P1 |
500 μL | Concentrated | 200μg/mL |
£323.00 £303.01 / 500µL Save £19.99 6% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
|
12639636
|
Epredia
MS-1352-P |
1 mL | Concentrated | 200μg/mL |
£429.00 £403.00 / 1mL Save £26.00 6% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
|
12619646
|
Epredia
MS-1352-R7 |
7 mL | Prediluted | Prediluted | |||||
|
12609646
|
Epredia
MS-1352-RQ |
12 mL | Ready-to-Use | Prediluted |
£349.00 £327.47 / 12mL Save £21.53 6% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Cytokeratin 7 is a basic cytokeratin which is found in most glandular and transitional epithelia but not in the stratified squamous epithelia. Keratin 7 is expressed in the epithelial cells of ovary, lung, and breast but not of colon, prostate, or gastrointestinal tract. Antibody to cytokeratin is useful in distinguishing ovarian carcinomas (keratin 7+) from colon carcinomas (keratin 7-).Host Species: Mouse
Clone: OV-TL 12/30
Isotype: IgG1/κ
Species Reactivity: Human. Does not react with rat. Others not known.
Epitope: Not determined
Immunogen: OTN II ovarian carcinoma cell line
Molecular Weight: 54 kDa
Positive Control: Ovarian CA
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
| Cytokeratin 7 Ab-2 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| IVD | |
| OTN II ovarian carcinoma cell line |
| OV-TL 12/30 | |
| Monoclonal | |
| Mouse | |
| Cytoskeletal Protein | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.