Learn More
Cytokeratin 7 Ab-2 Mouse Monoclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Cytokeratin 7 Ab-2, Mouse Monoclonal Antibody.
Brand: Epredia MS-1352-P
Description
Cytokeratin 7 is a basic cytokeratin which is found in most glandular and transitional epithelia but not in the stratified squamous epithelia. Keratin 7 is expressed in the epithelial cells of ovary, lung, and breast but not of colon, prostate, or gastrointestinal tract. Antibody to cytokeratin is useful in distinguishing ovarian carcinomas (keratin 7+) from colon carcinomas (keratin 7-).Host Species: Mouse
Clone: OV-TL 12/30
Isotype: IgG1/κ
Species Reactivity: Human. Does not react with rat. Others not known.
Epitope: Not determined
Immunogen: OTN II ovarian carcinoma cell line
Molecular Weight: 54 kDa
Positive Control: Ovarian CA
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
| Cytokeratin 7 Ab-2 | |
| Monoclonal | |
| 200μg/mL | |
| Mouse | |
| 1 mL | |
| Cytoskeletal Protein | |
| Human | |
| IgG2a κ |
| Immunohistochemistry (Paraffin) | |
| OV-TL 12/30 | |
| Unconjugated | |
| OTN II ovarian carcinoma cell line | |
| IVD | |
| Primary | |
| Concentrated |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.