missing translation for 'onlineSavingsMsg'
Learn More

Cytokeratin 7 Ab-2 Mouse Monoclonal Antibody, Epredia™

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Cytokeratin 7 Ab-2, Mouse Monoclonal Antibody.

Brand:  Epredia MS-1352-RQ

 View more versions of this product

Product Code. 12609646

  • ONLINE EXCLUSIVE PRICE
    £349.00
    £327.47 / 12mL
    Save £21.53
    6% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Cytokeratin 7 is a basic cytokeratin which is found in most glandular and transitional epithelia but not in the stratified squamous epithelia. Keratin 7 is expressed in the epithelial cells of ovary, lung, and breast but not of colon, prostate, or gastrointestinal tract. Antibody to cytokeratin is useful in distinguishing ovarian carcinomas (keratin 7+) from colon carcinomas (keratin 7-).

Host Species: Mouse

Clone: OV-TL 12/30

Isotype: IgG1/κ

Species Reactivity: Human. Does not react with rat. Others not known.

Epitope: Not determined

Immunogen: OTN II ovarian carcinoma cell line

Molecular Weight: 54 kDa

Positive Control: Ovarian CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cytokeratin 7 Ab-2
Monoclonal
Prediluted
Mouse
12 mL
Cytoskeletal Protein
Ready-to-Use
Immunohistochemistry (Paraffin)
OV-TL 12/30
Unconjugated
OTN II ovarian carcinoma cell line
IVD
Primary
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.