Learn More
Cyclin D3 Ab-1 Mouse Monoclonal Antibody, Epredia™
Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, immunoprecipitation and flow cytometry applications.
£138.00 - £481.44
Specifications
| Antigen | Cyclin D3 Ab-1 |
|---|---|
| Clone | DCS-22 |
| Applications | Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Form | Concentration | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Form | Concentration | Price | Quantity & Availability | |||
|
12690567
|
Epredia
MS-215-P0 |
100 μL | Concentrated | 200μg/mL | |||||
|
12610577
|
Epredia
MS-215-P1 |
500 μL | Concentrated | 200μg/mL |
£377.00 £354.31 / 500µL Save £22.69 6% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
|
12680567
|
Epredia
MS-215-P |
1 mL | Concentrated | 200μg/mL | |||||
|
12600577
|
Epredia
MS-215-P1ABX |
100 μL | Concentrated | 1mg/mL | |||||
|
12620577
|
Epredia
MS-215-PABX |
200 μL | Concentrated | 1mg/mL | |||||
|
12650577
|
Epredia
MS-215-R7 |
7 mL | Prediluted | — | |||||
Description
Cyclin D3 is a G1 cyclin closely related to cyclin D1 and D2. Cyclins D1, D2, and D3 form active kinase complexes in vivo with a distinct subset of cdks, particularly with cdk4 and cdk6.Host Species: Mouse
Clone: DCS-22
Isotype: IgG1/κ
Species Reactivity: Human, Monkey, Mouse and Rat. Others not known.
Epitope: Not determined
Immunogen: Purified human recombinant full-length cyclin D3 protein
Molecular Weight: 34 kDa
Positive Control: LS174T cells, Breast CA or tonsil
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Native only)
- Flow Cytometry
Note:
USA: RUO; Int'l: RUO
Specifications
| Cyclin D3 Ab-1 | |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Non-human Primate, Rat | |
| IgG2a κ |
| DCS-22 | |
| Monoclonal | |
| Mouse | |
| Cell Cycle | |
| Purified human recombinant full length cyclin D3 protein | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.