Learn More
Cyclin D3 Ab-1 Mouse Monoclonal Antibody, Epredia™
Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, immunoprecipitation and flow cytometry applications.
Brand: Epredia MS-215-PABX
Description
Cyclin D3 is a G1 cyclin closely related to cyclin D1 and D2. Cyclins D1, D2, and D3 form active kinase complexes in vivo with a distinct subset of cdks, particularly with cdk4 and cdk6.Host Species: Mouse
Clone: DCS-22
Isotype: IgG1/κ
Species Reactivity: Human, Monkey, Mouse and Rat. Others not known.
Epitope: Not determined
Immunogen: Purified human recombinant full-length cyclin D3 protein
Molecular Weight: 34 kDa
Positive Control: LS174T cells, Breast CA or tonsil
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Native only)
- Flow Cytometry
Note:
USA: RUO; Int'l: RUO
Specifications
| Cyclin D3 Ab-1 | |
| Monoclonal | |
| 1mg/mL | |
| Mouse | |
| 200 μL | |
| Cell Cycle | |
| Human, Mouse, Non-human Primate, Rat | |
| IgG2a κ |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| DCS-22 | |
| Unconjugated | |
| Purified human recombinant full length cyclin D3 protein | |
| RUO | |
| Primary | |
| Concentrated |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.