missing translation for 'onlineSavingsMsg'
Learn More

Cyclin D3 Ab-1 Mouse Monoclonal Antibody, Epredia™

Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, immunoprecipitation and flow cytometry applications.

Brand:  Epredia MS-215-P1

 View more versions of this product

Product Code. 12610577

  • ONLINE EXCLUSIVE PRICE
    £377.00
    £354.31 / 500µL
    Save £22.69
    6% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Cyclin D3 is a G1 cyclin closely related to cyclin D1 and D2. Cyclins D1, D2, and D3 form active kinase complexes in vivo with a distinct subset of cdks, particularly with cdk4 and cdk6.

Host Species: Mouse

Clone: DCS-22

Isotype: IgG1/κ

Species Reactivity: Human, Monkey, Mouse and Rat. Others not known.

Epitope: Not determined

Immunogen: Purified human recombinant full-length cyclin D3 protein

Molecular Weight: 34 kDa

Positive Control: LS174T cells, Breast CA or tonsil

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native only)
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cyclin D3 Ab-1
Monoclonal
200μg/mL
Mouse
500 μL
Cell Cycle
Human, Mouse, Non-human Primate, Rat
IgG2a κ
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
DCS-22
Unconjugated
Purified human recombinant full length cyclin D3 protein
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 500 UL CYCLIN D3 AB-1 MOUSE MAB (PURIFIED AB WITH BSA and Azide), Isotype: IgG1/k, Species reactivit y: Hu, Nhp, Ms, Rt, Application: IH, WB, IP, IF, FC 200µg/mL 0.5mL store at 2° to 8°C

Supplemental information
  • MIXTURE LIST-Contains : 5-chloro-2-methyl-3(2H)-isothiazolone, 2-methyl-3(2H)-isothiazolone, 5-Bromo-5-nitro-1,3-dioxane

SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.