missing translation for 'onlineSavingsMsg'
Learn More

CD68/Macrophage Marker Ab-3 Mouse Monoclonal Antibody, Epredia™

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ CD68/Macrophage Marker Ab-3, Mouse Monoclonal Antibody.

£113.22 - £382.50

Specifications

Antigen CD68/Macrophage Marker Ab-3
Clone KP1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 7
Product Code Brand Quantity Form Concentration Price Quantity & Availability  
Product Code Brand Quantity Form Concentration Price Quantity & Availability  
12641317
View Documents
Epredia
MS-397-P0
100 μL Concentrated 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12691317
View Documents
Epredia
MS-397-P1ABX
100 μL Concentrated 1mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12681317
View Documents
Epredia
MS-397-PABX
200 μL Concentrated 1mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12651317
View Documents
Epredia
MS-397-P1
500 μL Concentrated 200μg/mL
ONLINE EXCLUSIVE PRICE
£318.00
£297.59 / 500µL
Save £20.41
6% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12621317
View Documents
Epredia
MS-397-P
1 mL Concentrated 200μg/mL
ONLINE EXCLUSIVE PRICE
£401.00
£376.87 / 1mL
Save £24.13
6% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12601327
View Documents
Epredia
MS-397-R7
7 mL Prediluted
ONLINE EXCLUSIVE PRICE
£212.00
£187.72 / 7mL
Save £24.28
11% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12611327
View Documents
Epredia
MS-397-RQ
12 mL Supernatant This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

CD68 is expressed on macrophages and monocytes.

Host Species: Mouse

Clone: KP1

Isotype: IgG1/κ

Species Reactivity: Human, Monkey, Cat and Rat. Does not react with pig, dog and chicken.

Epitope: Not determined

Immunogen: Subcellular fraction of human alveolar macrophages

Molecular Weight: 110 kDa

Positive Control: Tonsil, lymph node, or spleen

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Specifications

CD68/Macrophage Marker Ab-3
Immunohistochemistry (Paraffin)
Unconjugated
IVD
Subcellular fraction of human alveolar macrophages
KP1
Monoclonal
Mouse
Hematopoietic
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.