Learn More
CD68/Macrophage Marker Ab-3 Mouse Monoclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ CD68/Macrophage Marker Ab-3, Mouse Monoclonal Antibody.
Brand: Epredia MS-397-P0
Description
CD68 is expressed on macrophages and monocytes.Host Species: Mouse
Clone: KP1
Isotype: IgG1/κ
Species Reactivity: Human, Monkey, Cat and Rat. Does not react with pig, dog and chicken.
Epitope: Not determined
Immunogen: Subcellular fraction of human alveolar macrophages
Molecular Weight: 110 kDa
Positive Control: Tonsil, lymph node, or spleen
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
| CD68/Macrophage Marker Ab-3 | |
| Monoclonal | |
| 200μg/mL | |
| Mouse | |
| 100 μL | |
| Hematopoietic | |
| Feline, Human, Non-human Primate, Rat | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| KP1 | |
| Unconjugated | |
| Subcellular fraction of human alveolar macrophages | |
| IVD | |
| Primary | |
| Concentrated |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.