missing translation for 'onlineSavingsMsg'
Learn More

CD68/Macrophage Marker Ab-3 Mouse Monoclonal Antibody, Epredia™

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ CD68/Macrophage Marker Ab-3, Mouse Monoclonal Antibody.

Brand:  Epredia MS-397-PABX

 View more versions of this product

Product Code. 12681317

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

CD68 is expressed on macrophages and monocytes.

Host Species: Mouse

Clone: KP1

Isotype: IgG1/κ

Species Reactivity: Human, Monkey, Cat and Rat. Does not react with pig, dog and chicken.

Epitope: Not determined

Immunogen: Subcellular fraction of human alveolar macrophages

Molecular Weight: 110 kDa

Positive Control: Tonsil, lymph node, or spleen

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD68/Macrophage Marker Ab-3
Monoclonal
1mg/mL
Mouse
200 μL
Hematopoietic
Feline, Human, Non-human Primate, Rat
IgG1 κ
Immunohistochemistry (Paraffin)
KP1
Unconjugated
Subcellular fraction of human alveolar macrophages
IVD
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.