missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Androgen Receptor Ab-1 Mouse Monoclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, immunoprecipitation, and gel supershift experiments.
£118.32 - £703.80
Specifications
Antigen | Androgen Receptor Ab-1 |
---|---|
Clone | AR 441 |
Classification | Monoclonal |
Host Species | Mouse |
Regulatory Status | RUO |
Product Code | Brand | Form | Conjugate | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Conjugate | Quantity | Price | Quantity & Availability | |||
12691427
|
Epredia
MS-443-P1 |
Concentrated | Unconjugated | 500 μL |
£375.00
500µL |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12671427
|
Epredia
MS-443-P |
Concentrated | Unconjugated | 1 mL |
£626.00
Each |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12681427
|
Epredia
MS-443-P0 |
Concentrated | Unconjugated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12621437
|
Epredia
MS-443-P1ABX |
Concentrated | Unconjugated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12611437
|
Epredia
MS-443-PABX |
Concentrated | Unconjugated | 200 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12651427
|
Epredia
MS-443-B0 |
Concentrated | Biotin | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12641427
|
Epredia
MS-443-B |
Concentrated | Biotin | 1 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12661437
|
Epredia
MS-443-R7 |
Prediluted | Unconjugated | 7 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12651437
|
Epredia
MS-443-RQ |
Ready-to-Use | Unconjugated | 12 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
The expression of AR is reportedly inversely correlated with histologic grade, i.e. well differentiated prostate tumors show higher expression than the poorly differentiated tumors. In prostate cancer, AR has been proposed as a marker of hormone-responsiveness.Host Species: Mouse
Clone: AR 441
Isotype: IgG1
Species Reactivity: Human and Dog. Does not react with mouse. Others not tested.
Epitope: aa 299-315
Immunogen: A synthetic peptide from human AR
Molecular Weight: 110kDa
Positive Control: LnCAp cells or prostate CA
Cellular Localization: Nuclear
Ab-1 is excellent for staining of formalin/paraffin tissues.
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Denatured verified)
- Gel Supershift
Note:
USA: RUO; Int'l: RUO
Specifications
Androgen Receptor Ab-1 | |
Monoclonal | |
RUO | |
A synthetic peptide from human AR |
AR 441 | |
Mouse | |
Steroids Hormones and Growth Factors | |
Primary |